accessibility menu, dialog, popup

UserName

CAS RN 98-44-2

NH2SHOOOSOHOO

CAS RN 98-44-2

IUPAC Name: 2-aminobenzene-1,4-disulfonic acid
Synonyms: 2-aminobenzene-1,4-disulfonic acid
Molecular Weight (g/mol): 253.24
Molecular Formula: C6H7NO6S2
InChi Key: LDCCBULMAFILCT-UHFFFAOYSA-N
SMILES: NC1=CC(=CC=C1S(O)(=O)=O)S(O)(=O)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)