missing translation for 'onlineSavingsMsg'
Learn More

Aniline-2,5-disulfonic acid, 95%

Product Code. 11497026
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11497026

Brand: Thermo Scientific Alfa Aesar B25350.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 98-44-2 | C6H7NO6S2 | 253.243 g/mol

Aniline-2,5-disulfonic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aniline-2,5-disulfonic acid is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-44-2
Molecular Formula C6H7NO6S2
Molecular Weight (g/mol) 253.243
MDL Number MFCD00035753
InChI Key LDCCBULMAFILCT-UHFFFAOYSA-N
Synonym 2-amino-1,4-benzenedisulfonic acid, aniline-2,5-disulfonic acid, 2,5-disulfoaniline, 4-sulfometanilic acid, 1,4-benzenedisulfonic acid, 2-amino, 2,5-disulfo-1-aminobenzene, 2-amino-1,4-disulfobenzene, 1-anilino-2,5-disulfonic acid, 2-amino-p-benzenedisulfonic acid, unii-5e2ot6t2ps
PubChem CID 60988
IUPAC Name 2-aminobenzene-1,4-disulfonic acid
SMILES C1=CC(=C(C=C1S(=O)(=O)O)N)S(=O)(=O)O

Specifications

Melting Point >300°C
Quantity 25 g
Formula Weight 253.25
Percent Purity 95%
Chemical Name or Material Aniline-2,5-disulfonic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.