missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LYST Antibody (3H1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00001130-M07
This item is not returnable.
View return policy
Description
LYST Monoclonal antibody specifically detects LYST in Human samples. It is validated for Western Blot, ELISA, ELISA
Specifications
| LYST | |
| Monoclonal | |
| Unconjugated | |
| NP_000072 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 1130 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2b κ |
| Western Blot, ELISA, Sandwich ELISA | |
| 3H1 | |
| In 1x PBS, pH 7.4 | |
| Beige homolog, Chediak-Higashi syndrome 1, CHSCHS1beige protein, lysosomal trafficking regulator, lysosomal-trafficking regulator | |
| LYST (NP_000072.2, 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MSTDSNSLAREFLTDVNRLCNAVVQRVEAREEEEEETHMATLGQYLVHGRGFLLLTKLNSIIDQALTCREELLTLLLSLLPLVWKIPVQEEKATDFNLPL | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction