missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD34 Antibody (MEC 14.7), Novus Biologicals™
Rat Monoclonal Antibody has been used in 22 publications
Brand: Novus Biologicals NB600-1071SS
This item is not returnable.
View return policy
Description
CD34 Monoclonal antibody specifically detects CD34 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
Specifications
| CD34 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:250, Flow Cytometry 1 ug per million cells, ELISA 1:100-1:2000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:100 | |
| CD34 | |
| murine transformed endothelioma cell line t-end. | |
| 0.025 mL | |
| Adaptive Immunity, Angiogenesis, B Cell Development and Differentiation Markers, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Hypoxia, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Tumor Suppressors | |
| 947 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| MEC 14.7 | |
| Unconjugated | |
| CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction