missing translation for 'onlineSavingsMsg'
Learn More

0.05 M (0.1 N) Sodium Arsenite Analytical Volumetric Solution (AVL), Reagecon
BIOPHARMA

Product Code. 11512952 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
2.5 L
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11512952 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11512952 Supplier Reagecon Supplier No. SA2005W

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fully factorized, high purity, stable product developed and tested for oxidation/reduction titrations

  • Tested to ± 0.1%
  • May be used for manual titrations or more commonly automatic titrations

Specifications

Packaging Amber glass Winchester bottle
Physical Form Liquid
Concentration 0.05 M
Technique Titration
Type Analytical Volumetric Solution
Quantity 2.5 L
compliance-icons
Product Identifier
  • SODIUM ARSENITE 0.1N ( 0.05M) Analytical Volumetric Solution
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 1A
Hazard Statement
  • H350-May cause cancer.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: diarsenic trioxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.