missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASPA Antibody (OTI3G5), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70216PCP
This item is not returnable.
View return policy
Description
ASPA Monoclonal antibody specifically detects ASPA in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| ASPA | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI3G5 | |
| Western Blot, Immunohistochemistry | |
| ACY-2, ACY2aminoacylase 2, Aminoacylase-2, ASPaminoacylase-2, aspartoacylase, aspartoacylase (aminoacylase 2, Canavan disease), EC 3.5.1.15 | |
| Human recombinant protein fragment corresponding to amino acids 77-313 of human ASPA(NP_000040) produced in E.coli. | |
| 0.1 mL | |
| Neuroscience | |
| 443 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction