missing translation for 'onlineSavingsMsg'
Learn More

BCOR Antibody (BCOR/1311) [Janelia Fluor™ 635], Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals (Bio-Techne)

Product Code. 30161075

  • 672.00 € / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

BCOR Monoclonal antibody specifically detects BCOR in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BCOR
Monoclonal
Janelia Fluor 635
ANOP2, BCL6 corepressor, BCL-6 corepressor, BCL6 co-repressor, BCL-6 interacting corepressor, BCoR, FLJ20285, FLJ38041, KIAA1575MGC71031, MAA2, MCOPS2, MGC131961
Recombinant fragment (around aa 100-400) of human BCOR protein (exact sequence is proprietary)
0.1 mL
Primary
Human
Purified
Immunohistochemistry (Paraffin)
BCOR/1311
50mM Sodium Borate
Mouse
Protein A or G purified
RUO
54880
Store at 4°C in the dark.
IgG2 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.