missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRTAP Antibody [Janelia Fluor™ 525], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-21432JF525
This item is not returnable.
View return policy
Description
CRTAP Polyclonal antibody specifically detects CRTAP in Human samples. It is validated for Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| CRTAP | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Janelia Fluor 525 | |
| cartilage associated protein, cartilage-associated protein, CASPLEPREL3, leprecan-like 3 | |
| Produced in rabbits immunized with E. coli-derived Human CRTAP fragment | |
| 0.1 mL | |
| Signal Transduction | |
| 10491 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction