missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FSH beta Antibody (FSHb/1062), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47734IR
This item is not returnable.
View return policy
Description
FSH beta Monoclonal antibody specifically detects FSH beta in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| FSH beta | |
| Monoclonal | |
| DyLight 755 | |
| follicle stimulating hormone, beta polypeptide, Follicle-stimulating hormone beta subunit, Follitropin beta chain, follitropin subunit beta, follitropin, beta chain, FSH-B, FSH-beta | |
| Recombinant full-length FSH beta sub-unit (Uniprot: P01225) | |
| 0.1 mL | |
| Cancer | |
| 2488 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| FSHb/1062 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction