Learn More
SCL/Tal1 Antibody (1E6), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-50285AF350
Product Code. 18702081
  • 467.00 € / 0.10mL
Description
SCL/Tal1 Monoclonal antibody specifically detects SCL/Tal1 in Zebrafish samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SCL/Tal1
Monoclonal
Alexa Fluor 350
BHLHA17, bHLHa17tal-1, Class A basic helix-loop-helix protein 17, SCLT-cell acute lymphocytic leukemia protein 1, Stem cell protein, TAL-1, T-cell acute lymphocytic leukemia 1, T-cell leukemia/lymphoma protein 5, TCL5tal-1 product
GST-SCL/Tal1 (N-terminal) fusion protein
0.1 mL
Primary
Zebrafish
Purified
Western Blot, Immunofluorescence
1E6
50 mM Sodium Borate
Mouse
Protein A purified
RUO
6886
Store at 4°C in the dark.
IgG1