missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF343 Antibody (PCRP-ZNF343-4F8), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24182MFV500
This item is not returnable.
View return policy
Description
ZNF343 Monoclonal antibody specifically detects ZNF343 in Human samples. It is validated for Flow Cytometry
Specifications
| ZNF343 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| FLJ3472, KOX21zinc finger protein 37a (KOX 21), zinc finger protein 37A, Zinc finger protein KOX21, ZNF37 | |
| Recombinant fragment of human ZNF343 protein | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| PCRP-ZNF343-4F8 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 79175 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction