missing translation for 'onlineSavingsMsg'
Learn More

0.25 mg/L C as Sucrose Total Organic Carbon (TOC) Standard for Sievers 900/M9 Analyzers, Reagecon
BIOPHARMA

Product Code. 14802211 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
35 mL
Unit Size:
35mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
14802211 35 mL 35mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 14802211 Supplier Reagecon Supplier No. RTOCS08
  • ONLINE EXCLUSIVE PRICE
    159.00 €
    150.03 € / 35mL
    Save 8.97 €
    5% Off
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extremely pure and specific product made from ultra-pure raw materials using a proprietary process.

  • Total Organic/Inorganic Carbon (TOC/TIC) Reference Standard, Total Organic/Inorganic Carbon (TOC/TIC) Reference Material.
  • Extremely high specification and purity.
  • Produced from ultra pure raw materials, including water that is prepared by a special proprietary process.
  • Presented in vials that are manufactured, cleaned and leached, by a special proprietary process specifically for the product.
  • Prepared and certified gravimetrically on a weight/volume basis to a specification of ± 2%.
  • Reagecon holds IEC/ISO 17025 accreditation for calibration of laboratory balances and pipettes (INAB Ref: 265C).
  • The resulting equipment calibration certificates are issued in accordance with the requirements of ISO/IEC 17025.
  • The concentration of these standards is verified using a high performance calibrated Total Organic Carbon Analyzer (TOC Instrument) using Reagecon’s ISO 17025 accredited test method (INAB Ref: 264T).
  • The calibration of the TOC instrument is completed using a high-purity ISO Guide 34 accredited TOC standard similar in TOC concentration value to this product.

Specifications

Description Accuracy: ± 2%, Matrix: water
Packaging TOC Glass Vial with Septa & Cap
Concentration 0.25 mg/L
Technique TOC Analysis
Quantity 35 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.