missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TER-119 Antibody (TER-119) [Janelia Fluor™ 635], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB1125JF635
This item is not returnable.
View return policy
Description
TER-119 Monoclonal antibody specifically detects TER-119 in Mouse samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunoprecipitation,Western Blot,CyTOF-reported
Specifications
| TER-119 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| Ly76, Ly-76, TER119 | |
| C57BL/6 mouse day-14 fetal liver cells | |
| 0.5 mL | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, CyTOF | |
| TER-119 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| 104231 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction