missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexafluoroisopropyl methacrylate, 99%, stab.

Product Code. 11479437
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11479437

Brand: Thermo Scientific Alfa Aesar B21447.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3063-94-3 | C7H6F6O2 | 236.113 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3063-94-3
Molecular Formula C7H6F6O2
Molecular Weight (g/mol) 236.113
MDL Number MFCD00040105
InChI Key FMQPBWHSNCRVQJ-UHFFFAOYSA-N
Synonym hexafluoroisopropyl methacrylate, 1,1,1,3,3,3-hexafluoroisopropyl methacrylate, 2-propenoic acid, 2-methyl-, 2,2,2-trifluoro-1-trifluoromethyl ethyl ester, methacrylic acid 1,1,1,3,3,3-hexafluoroisopropyl ester, 2,2,2-trifluoro-1-trifluoromethyl ethyl methacrylate, hexafluoro-2-propyl methacrylate, 1h-1-trifluoromethyl trifluoroethyl methacrylate, 1,1,1,3,3,3-hexafluoroprop-2-yl 2-methylprop-2-enoate, 1,1,1,3,3,3-hexafluoro-2-propanyl methacrylate, hfipma
PubChem CID 76469
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-yl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OC(C(F)(F)F)C(F)(F)F

Specifications

Density 1.302
Boiling Point 99°C
Flash Point 14°C (57°F)
Refractive Index 1.331
Quantity 5 g
UN Number UN1993
Formula Weight 236.12
Percent Purity 99%
Chemical Name or Material 1,1,1,3,3,3-Hexafluoroisopropyl methacrylate, stabilized

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.