missing translation for 'onlineSavingsMsg'
Learn More

1,1,3,3-Tetramethylbutyl isocyanate, 98%

Product Code. 11319485
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11319485 1 g 1g
11329485 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11319485 Supplier Thermo Scientific Alfa Aesar Supplier No. L18706.03
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1611-57-0 | C9H17NO | 155.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1611-57-0
Molecular Formula C9H17NO
Molecular Weight (g/mol) 155.24
MDL Number MFCD00209615
InChI Key XOHBENIMDRFUIH-UHFFFAOYSA-N
PubChem CID 407498
IUPAC Name 2-isocyanato-2,4,4-trimethylpentane
SMILES CC(C)(C)CC(C)(C)N=C=O

Specifications

Density 0.859
Boiling Point 170°C to 172°C
Flash Point 51°C (123°F)
Odor Pungent
Refractive Index 1.426
Quantity 1 g
UN Number UN3080
Sensitivity Moisture sensitive
Synonym 1,1,3,3-tetramethylbutyl isocyanate, tert-octyl isocyanate, 2-isocyanato-2,4,4-trimethyl-pentane, pentane, 2-isocyanato-2,4,4-trimethyl, acmc-1bnst, pentane,2-isocyanato-2,4,4-trimethyl, unavailable-see asdi_catno 500009647, 1,1,3,3-tetramethylbutyl isocyanate∼2,4,4-trimethyl-2-pentyl isocyanate
Formula Weight 155.24
Percent Purity 98%
Chemical Name or Material 1,1,3,3-Tetramethylbutyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.