missing translation for 'onlineSavingsMsg'
Learn More

1-(1,3-Benzodioxol-5-yl)-4,4,4-trifluorobutane-1,3-dione, 97%, Thermo Scientific™

Product Code. 10582843
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10582843 10 g 10g
10147082 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10582843 Supplier Thermo Scientific Maybridge Supplier No. KM10326EA
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 306935-39-7
Molecular Formula C11H7F3O4
Molecular Weight (g/mol) 260.168
MDL Number MFCD01570563
InChI Key FDLJUCOQPAGQNY-UHFFFAOYSA-N
Synonym 1-1,3-benzodioxol-5-yl-4,4,4-trifluorobutane-1,3-dione, 1-2h-1,3-benzodioxol-5-yl-4,4,4-trifluorobutane-1,3-dione, 3-3,4-methylenedioxy benzoyl-1,1,1-trifluoroacetone, 1-benzo d 1,3 dioxol-5-yl-4,4,4-trifluorobutane-1,3-dione, maybridge1_005379, 1,3-benzodioxol-5-yl-4,4,4-trifluorobutane-1,3-dion, 1,3-benzodioxol-5-yl-4,4,4-trifluorobutane-1,3-dione, 1,3-butanedione,1-1,3-benzodioxol-5-yl-4,4,4-trifluoro, 1-1,3-benzodioxol-5-yl-4,4,4-tri-fluorobutane-1,3-dione, 1-1,3-benzodioxole-5-yl-4,4,4-trifluorobutane-1,3-dione
PubChem CID 2775368
IUPAC Name 1-(1,3-benzodioxol-5-yl)-4,4,4-trifluorobutane-1,3-dione
SMILES C1OC2=C(O1)C=C(C=C2)C(=O)CC(=O)C(F)(F)F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 10 g
Formula Weight 260.17
Percent Purity 97%
Chemical Name or Material 1-(1,3-Benzodioxol-5-yl)-4,4,4-trifluorobutane-1,3-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.