missing translation for 'onlineSavingsMsg'
Learn More

1,1,4,4-Tetramethyl-1,2,3,4-tetrahydronaphthaline, Technical Grade, Thermo Scientific™

Product Code. 10575184
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10575184 100 g 100g
10672135 10 g 10g
10406003 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10575184 Supplier Thermo Scientific Maybridge Supplier No. TL00201FA
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 6683-46-1
Molecular Formula C14H20
Molecular Weight (g/mol) 188.31
MDL Number MFCD00052728
InChI Key CCQKWSZYTOCEIB-UHFFFAOYSA-N
PubChem CID 81186
IUPAC Name 1,1,4,4-tetramethyl-2,3-dihydronaphthalene
SMILES CC1(C)CCC(C)(C)C2=CC=CC=C12

Specifications

Assay Percent Range 90%
Packaging Amber glass bottle
Quantity 100 g
Synonym 1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphthalene, 1,2,3,4-tetrahydro-1,1,4,4-tetramethylnaphthalene, 1,1,4,4-tetramethyltetralin, naphthalene, 1,2,3,4-tetrahydro-1,1,4,4-tetramethyl, naphthalene,1,2,3,4-tetrahydro-1,1,4,4-tetramethyl, 1,2,3,4-tetrahydro-1,1,4,4tetramethylnaphthalene, 1,1,4,4-tetramethyl-1,2,3,4-tetrahydro naphthalene, 1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphthaline, 1,1,4,4-tetramethyl1,2,3,4-tetrahydro naphthalene, 1,2,3,4-tetrahydro-1,1,4,4-tetramethyl-naphthalene
Formula Weight 188.31
Percent Purity 90%
Chemical Name or Material 1,1,4,4-Tetramethyl-1,2,3,4-tetrahydronaphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.