missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dichlorodimethyl ether, 97%

Product Code. 10306420 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10306420 10 g Glass bottle 10g
10209463 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10306420 Supplier Thermo Scientific Acros Supplier No. 113470100
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4885-02-3 | C2H4Cl2O | 114.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4885-02-3
Molecular Formula C2H4Cl2O
Molecular Weight (g/mol) 114.96
MDL Number MFCD00000828
InChI Key GRTGGSXWHGKRSB-UHFFFAOYSA-N
SMILES COC(Cl)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2700g/mL
Boiling Point 85.0°C
Flash Point 22°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula Cl2CHOCH3
Refractive Index 1.4295 to 1.4325
Quantity 10 g
Fieser 01,220; 02,120; 05,200; 06,171; 07,98; 08,154
Specific Gravity 1.27
Synonym dichloromethyl methyl ether, 1,1-dichlorodimethyl ether, dichloro methoxy methane, methane, dichloromethoxy, ether, dichloromethyl methyl, alpha,alpha-dichloromethyl methyl ether, methoxydichloromethane, 1,1-dichloromethyl methyl ether, alpha,alpha-dichloromethyl ether, dichloromethylmethylether
IUPAC Name dichloro(methoxy)methane
PubChem CID 21004
Formula Weight 114.96
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1, 1-Dichlorodimethyl ether
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,1-Dichlorodimethyl ether
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • EUH018-In use, may form flammable/explosive vapour-air mixture.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.