missing translation for 'onlineSavingsMsg'
Learn More

1,1-Diphenyl-2-propyn-1-ol, 99%

Product Code. 11429407
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
11429407 5 g 5g
11439407 25 g 25g
2 options
This item is not returnable. View return policy

Product Code. 11429407

Brand: Thermo Scientific Alfa Aesar B21428.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3923-52-2 | C15H12O | 208.26 g/mol

1,1-Diphenyl-2-propyn-1-ol is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1-Diphenyl-2-propyn-1-ol is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used in medicine.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3923-52-2
Molecular Formula C15H12O
Molecular Weight (g/mol) 208.26
MDL Number MFCD00041570
InChI Key SMCLTAARQYTXLW-UHFFFAOYSA-N
Synonym 1,1-diphenyl-2-propyn-1-ol, diphenylethinylcarbinol, ethynyldiphenylmethanol, 1,1-diphenylpropargyl alcohol, diphenylethynylcarbinol, 3,3-diphenyl-3-hydroxy-1-propyne, benzenemethanol, .alpha.-ethynyl-.alpha.-phenyl, 2-propyn-1-ol, 1,1-diphenyl, benzenemethanol, alpha-ethynyl-alpha-phenyl, benzenemethanol, a-ethynyl-a-phenyl
PubChem CID 92976
IUPAC Name 1,1-diphenylprop-2-yn-1-ol
SMILES OC(C#C)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 44°C to 48°C
Boiling Point 179°C to 180°C (18 mmHg)
Flash Point 113°C (235°F)
Quantity 5 g
Beilstein 514549
Solubility Information Insoluble in water.
Formula Weight 208.26
Percent Purity 99%
Chemical Name or Material 1,1-Diphenyl-2-propyn-1-ol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.