missing translation for 'onlineSavingsMsg'
Learn More

1,1-Diphenylethylene, 98%

Product Code. 11457077
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
250 g
Unit Size:
100g
10g
250g
25g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11457077 10 g 10g
11404250 25 g 25g
11414250 100 g 100g
11467077 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11457077 Supplier Thermo Scientific Alfa Aesar Supplier No. A14434.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1,1-Diphenylethylene, 98%, C14H12, CAS Number-530-48-3, unii-bx0l5b6lll, 1,1-diphenylethene, 1,1-diphenylethylene, 1-phenylethenyl benzene, 1-phenylvinyl benzene, ethene-1,1-diyldibenzene, bx0l5b6lll, .alpha.-phenylstyrene, benzene, 1,1'-ethenylidenebis, as-diphenylethylene, 250g, 1099062 | CAS: 530-48-3 | C14H12 | 180.25 g/mol

1,1-Diphenylethylene is used in the preparation of 2-chloro-1,1-diphenyl-ethene by reacting with benzeneseleninyl chloride and aluminum(III) chloride as reagents. It acts as an intermediate in organic synthesis as well as in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1-Diphenylethylene is used in the preparation of 2-chloro-1,1-diphenyl-ethene by reacting with benzeneseleninyl chloride and aluminum(III) chloride as reagents. It acts as an intermediate in organic synthesis as well as in pharmaceuticals.

Solubility
Miscible with methanol, chloroform and ether. Insoluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 530-48-3
Molecular Formula C14H12
Molecular Weight (g/mol) 180.25
MDL Number MFCD00008583
InChI Key ZMYIIHDQURVDRB-UHFFFAOYSA-N
Synonym 1,1-diphenylethylene, ethene-1,1-diyldibenzene, 1,1-diphenylethene, benzene, 1,1'-ethenylidenebis, 1-phenylethenyl benzene, unii-bx0l5b6lll, as-diphenylethylene, .alpha.-phenylstyrene, 1-phenylvinyl benzene, bx0l5b6lll
PubChem CID 10740
IUPAC Name 1-phenylethenylbenzene
SMILES C=C(C1=CC=CC=C1)C2=CC=CC=C2

Specifications

Melting Point 5°C to 7°C
Density 1.023
Boiling Point 270°C to 271°C
Flash Point 136°C (276°F)
Refractive Index 1.6065
Quantity 10 g
Beilstein 1099062
Merck Index 14,3323
Solubility Information Miscible with methanol,chloroform and ether. Insoluble in water.
Formula Weight 180.25
Percent Purity 98%
Chemical Name or Material 1,1-Diphenylethylene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.