missing translation for 'onlineSavingsMsg'
Learn More

1-(1-Hydroxy-4-Methyl-2-pHenylazo)-2-NapHthol-4-Sulfonic Acid, Pure, Metal Indicator, Fisher Chemical

Product Code. 10627222 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass Jar
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10627222 5 g Glass Jar 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10627222 Supplier Fisher Chemical Supplier No. H/2350/44
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 3147-14-6
Melting Point 300°C
Color Black
Packaging Glass Jar
Molecular Formula C17H14N2O5S
MDL Number 11656
Quantity 5 g
Synonym calmagite, calmagite, indicator grade, 1-naphthalenesulfonic acid, 3-hydroxy-4-2-hydroxy-5-methylphenyl azo, 3-hydroxy-4-6-hydroxy-m-tolyl azo-1-naphthalenesulfonic acid, calmagite indicator grade, 1-naphthalenesulfonic acid, 3-hydroxy-4-2-2-hydroxy-5-methylphenyl diazenyl, vbrnloqcbcpphl-uhfffaoysa-n, calmagite metalindicatorforca,mgetc., 3-hydroxy-4-2-hydroxy-5-methyl-phenylazo-1-naphthalenesulfonic acid, e-3-hydroxy-4-2-hydroxy-5-methylphenyl diazenyl naphthalene-1-sulfonic acid
InChI Key ASFVMSDYPYMUFL-UHFFFAOYSA-N
SMILES CC1=CC(=C(C=C1)O)NN=C2C3=CC=CC=C3C(=CC2=O)S(=O)(=O)O
IUPAC Name 4-[(2-hydroxy-5-methylphenyl)hydrazinylidene]-3-oxonaphthalene-1-sulfonic acid
Molecular Weight (g/mol) 358.368
PubChem CID 6364506
Formula Weight 358.38g/mol
Physical Form Powder
Show More Show Less
compliance-icons
Product Identifier
  • Calmagite
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.