missing translation for 'onlineSavingsMsg'
Learn More

1-[2-(2,4-Difluorophenyl)-2,3-epoxypropyl]-1H-1,2,4-triazole, TRC

Product Code. 30010222
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010222 1 g 1g
30005354 500 mg 500mg
30008127 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010222 Supplier Toronto Research Chemicals Supplier No. TRCD4460451G
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86386-76-7
Molecular Formula C11 H9 F2 N3 O
Molecular Weight (g/mol) 237.21
Synonym 1H-1,2,4-Triazole, 1-[[2-(2,4-difluorophenyl)-2-oxiranyl]methyl]-, 1H-1,2,4-Triazole, 1-[[2-(2,4-difluorophenyl)oxiranyl]methyl]- (9CI), 1-[[2-(2,4-Difluorophenyl)-2-oxiranyl]methyl]-1H-1,2,4-triazole, 1-[2-(2,4-Difluorophenyl)-2,3-epoxypropyl]-1H-1,2,4-triazole, 1-[[2-(2,4-Difluorophenyl)oxiranyl]methyl]-1H-1,2,4-triazole, 2-(2,4-Difluorophenyl)-2,3-epoxy-1-(1H-1,2,4-triazol-1-yl)propane, 2-(2,4-Difluorophenyl)-2-[(1H-1,2,4-triazol-1-yl)methyl]oxirane, 1-[[(2RS)-2-(2,4-Difluorophenyl)oxiran-2-yl]methyl]-1H-1,2,4-triazole
IUPAC Name 1-[[2-(2,4-difluorophenyl)oxiran-2-yl]methyl]-1,2,4-triazole
SMILES Fc1ccc(c(F)c1)C2(Cn3cncn3)CO2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C11H9F2N3O/c12-8-1-2-9(10(13)3-8)11(5-17-11)4-16-7-14-6-15-16/h1-3,6-7H,4-5H2
Formula Weight 237.0714
Chemical Name or Material 1-[2-(2,4-Difluorophenyl)-2,3-epoxypropyl]-1H-1,2,4-triazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.