missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-1H-Triazole, 97%

Product Code. 10766062 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10766062 1 g Glass bottle 1g
10766252 5 g Glass bottle 5g
11916581 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10766062 Supplier Thermo Scientific Acros Supplier No. 345480010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 288-36-8 | C2H3N3 | 69.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 288-36-8
Molecular Formula C2H3N3
Molecular Weight (g/mol) 69.07
InChI Key QWENRTYMTSOGBR-UHFFFAOYSA-N
Synonym 1h-1,2,3-triazole, 1,2,3-triazole, 2h-1,2,3-triazole, 1,2,3-1h-triazole, triazole, triazol, osotriazole, v-triazole, 1,2,3-triazol, 1,2,3 triazole
PubChem CID 67516
ChEBI CHEBI:35565
IUPAC Name 2H-triazole
SMILES C1=NNN=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 23.0°C to 25.0°C
Color Colorless to Yellow
Density 1.1920g/mL
Boiling Point 203.0°C (752.0 mmHg)
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4930 to 1.4960
Quantity 1 g
Specific Gravity 1.192
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetone,diethyl ether,insoluble in n-pentane
Formula Weight 69.07
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1, 2, 3-1H-Triazole
Show More Show Less
compliance-icons
Product Identifier
  • 1,2,3-1H-Triazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.