missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4,6-Penta-O-galloyl-Beta-D-glucopyranose >90%, TRC

Product Code. 30133411
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133411 25 mg 25mg
30006068 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133411 Supplier Toronto Research Chemicals Supplier No. TRCP27045025MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14937-32-7
Molecular Formula C41 H32 O26
Molecular Weight (g/mol) 940.68
Synonym β-D-Glucopyranose, 1,2,3,4,6-pentakis(3,4,5-trihydroxybenzoate), Gallic acid, pentaester with β-D-glucopyranose (6CI), Glucopyranose, pentagallate, β-D- (8CI), β-D-Glucopyranose, pentakis(3,4,5-trihydroxybenzoate) (9CI), 1,2,3,4,6-Penta-O-galloyl-β-D-glucopyranose, 1,2,3,4,6-Penta-O-galloyl-β-D-glucopyranoside, 1,2,3,4,6-Penta-O-galloyl-β-D-glucose, 1,2,3,4,6-Penta-O-galloyl-β-glucopyranose, 1,2,3,4,6-Penta-O-galloylglucose, 1,2,3,4,6-Pentagalloyl-β-D-glucopyranose, 1,2,3,4,6-Pentagalloyl-β-D-glucopyranoside, 1,2,3,4,6-Pentagalloyl-β-D-glucose, Gallotannin 15, Penta-O-galloyl-β-D-glucopyranose, Penta-O-galloyl-β-D-glucose, Penta-O-galloylglucose, Pentagalloyl-D-glucose, β-1,2,3,4,6-Pentagalloylglucose
IUPAC Name [(2R,3R,4S,5R,6S)-3,4,5,6-tetrakis[(3,4,5-trihydroxybenzoyl)oxy]oxan-2-yl]methyl 3,4,5-trihydroxybenzoate
SMILES Oc1cc(cc(O)c1O)C(=O)OC[C@H]2O[C@@H](OC(=O)c3cc(O)c(O)c(O)c3)[C@H](OC(=O)c4cc(O)c(O)c(O)c4)[C@@H](OC(=O)c5cc(O)c(O)c(O)c5)[C@@H]2OC(=O)c6cc(O)c(O)c(O)c6

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C41H32O26/c42-17-1-12(2-18(43)28(17)52)36(57)62-11-27-33(64-37(58)13-3-19(44)29(53)20(45)4-13)34(65-38(59)14-5-21(46)30(54)22(47)6-14)35(66-39(60)15-7-23(48)31(55)24(49)8-15)41(63-27)67-40(61)16-9-25(50)32(56)26(51)10-16/h1-10,27,33-35,41-56H,11H2/t27-,33-,34+,35-,41+/m1/s1
Formula Weight 940.1182
Chemical Name or Material 1,2,3,4,6-Penta-O-galloyl-beta-D-glucopyranose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.