missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 1,2:3,4-Di-O-isopropylidene-D-galactopyranose, 97%

Product Code. 10204670 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10204670 25 g Glass bottle 25g
10040810 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10204670 Supplier Thermo Scientific Chemicals Supplier No. 115240250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4064-06-6 | C12H20O6 | 260.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4064-06-6
Molecular Formula C12H20O6
Molecular Weight (g/mol) 260.28
MDL Number MFCD00063225
InChI Key POORJMIIHXHXAV-UHFFFAOYNA-N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 120.0°C to 122.0°C
Color Yellow
Density 1.1400g/mL
Boiling Point 117.0°C (0.1 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4668 to 1.4688
Quantity 25 g
Specific Gravity 1.14
Specific Rotation Condition − 57.50 (20.00°C c=3,CHCl3)
Specific Rotation -57.5
Synonym n-4-methylphenyl-2,2-diphenylacetamide, 2,2-diphenyl-n-p-tolylacetamide, cbmicro_030789, n-p-tolyl-2,2-diphenylacetamide, 2,2-diphenyl-n-p-tolyl acetamide, nbeta-z-nin-boc-l-beta-homotryptophan, 2,2-diphenyl-n-p-tolyl-acetamide, n-4-methylphenyl-2,2-diphenyl-acetamide, n∼1∼-4-methylphenyl-2,2-diphenylacetamide
SMILES CC1(C)OC2OC(CO)C3OC(C)(C)OC3C2O1
IUPAC Name {4,4,11,11-tetramethyl-3,5,7,10,12-pentaoxatricyclo[7.3.0.0²,⁶]dodecan-8-yl}methanol
PubChem CID 344833
Formula Weight 260.28
Percent Purity 97%
Physical Form Viscous Paste
Chemical Name or Material 1, 2:3, 4-Di-O-isopropylidene-D-galactopyranose, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.