missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetrahydronaphthalene, 97%

Product Code. 11437167
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11437167 1000 g 1000g
11447167 250 g 250g
11457167 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11437167 Supplier Thermo Scientific Alfa Aesar Supplier No. A14962.0B
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 119-64-2 | C10H12 | 132.21 g/mol

1,2,3,4-Tetrahydronaphthalene, is used as an intermediate for organic synthesis, solvent. It is used as a solvent.It is also used for the laboratory synthesis of dry HBr gas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,3,4-Tetrahydronaphthalene, is used as an intermediate for organic synthesis, solvent. It is used as a solvent.It is also used for the laboratory synthesis of dry HBr gas.

Solubility
It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. Air and light sensitive. Heat sensitive. May darken on storage. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 119-64-2
Molecular Formula C10H12
Molecular Weight (g/mol) 132.21
MDL Number MFCD00001733
InChI Key CXWXQJXEFPUFDZ-UHFFFAOYSA-N
Synonym tetralin, benzocyclohexane, tetrahydronaphthalene, bacticin, tetraline, tetranap, naphthalene, 1,2,3,4-tetrahydro, tetralina, naphthalene 1,2,3,4-tetrahydride, tetralene
PubChem CID 8404
ChEBI CHEBI:35008
IUPAC Name 1,2,3,4-tetrahydronaphthalene
SMILES C1CCC2=CC=CC=C2C1

Spécification

Melting Point -36°C
Density 0.966
Boiling Point 206°C to 207°C
Flash Point 77°C (171°F)
Refractive Index 1.54
Quantity 1000 g
UN Number UN3082
Beilstein 1446407
Sensitivity Air Sensitive
Merck Index 14,9221
Solubility Information It is insoluble in water.
Formula Weight 132.21
Percent Purity 97%
Chemical Name or Material 1,2,3,4-Tetrahydronaphthalene
Afficher plus Afficher moins
compliance-icons compliance-icons
Identifiant produit
  • 1,2,3,4-Tetrahydronaphthalene
Mention d'avertissement
  • Warning
Catégorie de danger
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Mention de danger
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH019-May form explosive peroxides.
Conseils de prudence
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.
Informations supplémentaires
  • MIXTURE LIST-Contain: 1,2,3,4-Tetrahydronaphthalene

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.