missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Trifluorobenzene, 98+%

Product Code. 11499547
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11499547 1 g 1g
11409557 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499547 Supplier Thermo Scientific Alfa Aesar Supplier No. B22055.03
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1489-53-8 | C6H3F3 | 132.085 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1489-53-8
Molecular Formula C6H3F3
Molecular Weight (g/mol) 132.085
MDL Number MFCD00012013
InChI Key AJKNNUJQFALRIK-UHFFFAOYSA-N
Synonym trifluorobenzene, benzene, trifluoro, benzene, 1,2,3-trifluoro, 3,4,5-trifluoro-benzene, pubchem1067, 1,2,3-fluorobenzene, 1,2,3-trifluorobenze, 1,2,3-trifluorbenzen, 1,2,6-trifluorobenzene, 2,3,4-trifluorobenzene
PubChem CID 15147
IUPAC Name 1,2,3-trifluorobenzene
SMILES C1=CC(=C(C(=C1)F)F)F

Specifications

Density 1.28
Boiling Point 94°C to 95°C
Flash Point −3°C (26°F)
Refractive Index 1.422
Quantity 1 g
UN Number UN2387
Beilstein 1862388
Formula Weight 132.09
Percent Purity ≥98%
Chemical Name or Material 1,2,3-Trifluorobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.