missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Trimethoxybenzene, 98+%

Product Code. 11431968
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11431968

Brand: Thermo Scientific Alfa Aesar A14572.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 634-36-6 | C9H12O3 | 168.192 g/mol

1,2,3-Trimethoxybenzene was used to study the effect of solvent on photo induced electron-transfer reactions, on condensation with 2,4-diamino-5-(hydroxymethyl)pyrimidine yields 2,4-diamino-5-(2,3,4-trimethoxybenzyl)pyrimidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,3-Trimethoxybenzene was used to study the effect of solvent on photo induced electron-transfer reactions, on condensation with 2,4-diamino-5-(hydroxymethyl)pyrimidine yields 2,4-diamino-5-(2,3,4-trimethoxybenzyl)pyrimidine.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 634-36-6
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.192
MDL Number MFCD00008358
InChI Key CRUILBNAQILVHZ-UHFFFAOYSA-N
Synonym methylsyringol, pyrogallol trimethyl ether, trimethoxybenzene, benzene, 1,2,3-trimethoxy, tri-o-methylpyrogallol, benzene, trimethoxy, unii-mre1o894fg, 1,2,3-trimethoxy benzene, mre1o894fg, 3,4,5-trimethoxybenzene
PubChem CID 12462
ChEBI CHEBI:86529
IUPAC Name 1,2,3-trimethoxybenzene
SMILES COC1=C(C(=CC=C1)OC)OC

Specifications

Melting Point 43°C to 45°C
Density 1.112
Boiling Point 241°C
Flash Point 112°C (233°F)
Quantity 100 g
Beilstein 1910422
Solubility Information Insoluble in water.
Formula Weight 168.19
Percent Purity ≥98%
Chemical Name or Material 1,2,3-Trimethoxybenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.