missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Triazin-3-amine, TRC

Product Code. 30450631
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30450631 10 mg 10mg
30453618 100 mg -
30451446 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30450631 Supplier Toronto Research Chemicals Supplier No. TRCT79526310MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1120-99-6
Molecular Formula C3H4N4
Molecular Weight (g/mol) 96.09
Synonym 3-Amino-1,2,4-triazine, 3-Amino-as-triazine, NSC 203233, [1,2,4]Triazin-3-ylamine
IUPAC Name 4H-1,2,4-triazin-3-imine
SMILES N=C1NC=CN=N1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C3H4N4/c4-3-5-1-2-6-7-3/h1-2H,(H2,4,5,7)
Formula Weight 96.0436
Chemical Name or Material 1,2,4-Triazin-3-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.