missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Trimethoxybenzene, 98%

Product Code. 11468723
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11468723 50 g 50g
11458723 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11468723 Supplier Thermo Scientific Alfa Aesar Supplier No. A10483.18
 Restricted Item
Estimated Shipment: 11-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 135-77-3 | C9H12O3 | 168.192 g/mol

1,2,4-Trimethoxybenzene is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,4-Trimethoxybenzene is used as pharmaceutical intermediates.

Solubility
Not miscible in water.

Notes
Store away from incompatible materials such as oxidizing agents, acids and bases. Use only in a chemical fumehood.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135-77-3
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.192
MDL Number MFCD00008360
InChI Key AGIQIOSHSMJYJP-UHFFFAOYSA-N
Synonym benzene, 1,2,4-trimethoxy, unii-3ku3wd07ss, epa pesticide chemical code 040515, 3ku3wd07ss, hydroxyhydroquinone trimethyl ether, 2,4,5-trimethoxybenzene, acmc-1br3s, 1,3,4-trimethoxy benzene, dsstox_cid_15163, dsstox_rid_79243
PubChem CID 67284
IUPAC Name 1,2,4-trimethoxybenzene
SMILES COC1=CC(=C(C=C1)OC)OC

Spezifikation

Density 1.128
Boiling Point 247°C
Flash Point >110°C (230°F)
Refractive Index 1.533
Quantity 50 g
Beilstein 2047579
Merck Index 14,1073
Solubility Information Not miscible in water.
Formula Weight 168.19
Percent Purity 98%
Chemical Name or Material 1,2,4-Trimethoxybenzene
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.