missing translation for 'onlineSavingsMsg'
Learn More

1,2,7,8-Diepoxyoctane, 97%, Thermo Scientific Chemicals

Product Code. 11376986
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11376986 10 g 10g
11386986 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11376986 Supplier Thermo Scientific Alfa Aesar Supplier No. L06768.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1,2,7,8-Diepoxyoctane is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,7,8-Diepoxyoctane is used as an organic chemical synthesis intermediate.

Solubility
Soluble in water (partly miscible), acetone and heptane.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2426-07-5
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00005155
InChI Key LFKLPJRVSHJZPL-UHFFFAOYNA-N
Synonym 1,2,7,8-diepoxyoctane, 1,2:7,8-diepoxyoctane, 1,7-octadiene diepoxide, oxirane, 2,2'-1,4-butanediyl bis, 1,2-epoxy-7,8-epoxyoctane, 2,2'-1,4-butanediyl bisoxirane, 1,4-di oxiran-2-yl butane, octane, 1,2:7,8-diepoxy, 2-4-oxiran-2-yl butyl oxirane, ccris 963
PubChem CID 17048
ChEBI CHEBI:23705
IUPAC Name 2-[4-(oxiran-2-yl)butyl]oxirane
SMILES C1C(O1)CCCCC2CO2

Specifications

Density 0.997
Boiling Point 238°C to 240°C
Flash Point 97°C (206°F)
Refractive Index 1.445
Quantity 10 g
UN Number UN2922
Beilstein 104873
Solubility Information Soluble in water (partly miscible),acetone and heptane.
Formula Weight 142.2
Percent Purity 97%
Chemical Name or Material 1,2,7,8-Diepoxyoctane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.