missing translation for 'onlineSavingsMsg'
Learn More

1,2-Benzanthracene, 99%

Product Code. 10628124 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10628124 250 mg Glass bottle 250mg
10569800 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10628124 Supplier Thermo Scientific Acros Supplier No. 105252500
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 56-55-3 | C18H12 | 228.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-55-3
Molecular Formula C18H12
Molecular Weight (g/mol) 228.29
MDL Number MFCD00003599
InChI Key DXBHBZVCASKNBY-UHFFFAOYSA-N
Synonym benz a anthracene, tetraphene, 1,2-benzanthracene, benzanthrene, benzo a anthracene, benzanthracene, naphthanthracene, 2,3-benzphenanthrene, benzoanthracene
PubChem CID 5954
ChEBI CHEBI:51348
IUPAC Name benzo[a]anthracene
SMILES C1=CC=C2C=C3C(C=CC4=CC=CC=C34)=CC2=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 158°C to 161°C
Color Beige to Green-Yellow
Boiling Point 437.6°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 250 mg
Beilstein 05,718
Merck Index 15,1064
Solubility Information Solubility in water: insoluble in water. Other solubilities: difficulty soluble in boiling alcohol,soluble in most other common organic solvents
Formula Weight 228.29
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 1, 2-Benzanthracene, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,2-Benzanthracene
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.