missing translation for 'onlineSavingsMsg'
Learn More

1,2-Benzenedimethanol, 97%

Product Code. 10562011 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10562011 2.5 g Glass bottle 2.5g
10521564 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10562011 Supplier Thermo Scientific Supplier No. 180410025
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 612-14-6 | C8H10O2 | 138.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 612-14-6
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00004626
InChI Key XMUZQOKACOLCSS-UHFFFAOYSA-N
Synonym 1,2-benzenedimethanol, 1,2-phenylenedimethanol, phthalyl alcohol, 1,2-dihydroxymethylbenzene, o-xylene-alpha,alpha'-diol, 2-hydroxymethyl phenyl methanol, benzenedimethanol, o-xylylene glycol, 1,2-di hydroxymethyl benzene, 2-hydroxymethylbenzenemethanol
PubChem CID 69153
IUPAC Name [2-(hydroxymethyl)phenyl]methanol
SMILES OCC1=CC=CC=C1CO

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 61.0°C to 64.0°C
Color White to Yellow
Boiling Point 291.0°C
Flash Point 145°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6H4(CH2OH)2
Quantity 2.5 g
Beilstein 06, 910
Fieser 16,18; 12,563; 13,341
Solubility Information Solubility in water: soluble
Formula Weight 138.17
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1, 2-Benzenedimethanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.