missing translation for 'onlineSavingsMsg'
Learn More

1,2-Benzisoxazole, 95%

Product Code. 11461007
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11461007 1 g 1g
11471007 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11461007 Supplier Thermo Scientific Alfa Aesar Supplier No. B23400.03
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 271-95-4 | C7H5NO | 119.123 g/mol

1,2-Benzisoxazole derivative zonisamide is a novel antiepileptic drug and is effective for the treatment of partial seizures. 1,2-Benzisoxazole is a potential substrates of rabbit liver aldehyde oxidase. It is used to produce 5-nitro-benzo[d]isoxazole. It is involved in manufacturing medicines ( antiepileptics and antipsychotics).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water. 4.1 mg/mL in water at 25°C

Notes
Light Sensitive. Store at 4°C. Store in the dark. Incompatible with light and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 271-95-4
Molecular Formula C7H5NO
Molecular Weight (g/mol) 119.123
MDL Number MFCD00005852
InChI Key KTZQTRPPVKQPFO-UHFFFAOYSA-N
Synonym 1,2-benzisoxazole, benzo d isoxazole, indoxazene, benzisoxazole, benz d isoxazole, 4,5-benzisoxazole, 1-oxa-2-aza-1h-indene, benzoisoxazole, indoxazine, isoindoxazine
PubChem CID 71073
ChEBI CHEBI:51554
IUPAC Name 1,2-benzoxazole
SMILES C1=CC=C2C(=C1)C=NO2

Specifications

Density 1.169
Boiling Point 90°C to 92°C (15 mmHg)
Flash Point 86°C (186°F)
Refractive Index 1.562
Quantity 1 g
Beilstein 2154
Sensitivity Light sensitive
Solubility Information Soluble in water. 4.1mg/mL in water at 25°C
Formula Weight 119.12
Percent Purity 97%
Chemical Name or Material 1,2-Benzisoxazole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.