missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(2-nitrophenoxy)ethane, 98%, Thermo Scientific™

Product Code. 11490277
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11490277

Brand: Thermo Scientific Alfa Aesar B21692.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51661-19-9
Molecular Formula C14H12N2O6
Molecular Weight (g/mol) 304.26
MDL Number MFCD00024226
InChI Key DEHTVRKGDUCXRF-UHFFFAOYSA-N
Synonym 1,2-bis o-nitrophenoxy ethane, 1,2-bis 2-nitrophenoxy ethane, 1-nitro-2-2-2-nitrophenoxy ethoxy benzene, ethylene glycol bis 2-nitrophenyl ether, 1,1'-ethane-1,2-diylbis oxy bis 2-nitrobenzene, bis 2-nitrophenoxy ethane, acmc-20a59z, 1,2-di o-nitrophenoxy ethane, 2,2'-ethylenedioxy bis 1-nitrobenzene, 2-nitro-1-2-2-nitrophenoxy ethoxy benzene
PubChem CID 394580
IUPAC Name 1-nitro-2-[2-(2-nitrophenoxy)ethoxy]benzene
SMILES [O-][N+](=O)C1=CC=CC=C1OCCOC1=CC=CC=C1[N+]([O-])=O

Specifications

Melting Point 167°C to 170°C
Quantity 250 g
Formula Weight 304.26
Percent Purity 98%
Chemical Name or Material 1,2-Bis(2-nitrophenoxy)ethane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.