missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(methanesulfonamido)benzene, 97%

Product Code. 11458677
Change view
Click to view available options
Quantity:
2.5 g
10 g
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11458677 2.5 g 2.5g
11468677 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11458677 Supplier Thermo Scientific Alfa Aesar Supplier No. A19830.88
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 7596-80-7 | C8H12N2O4S2 | 264.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7596-80-7
Molecular Formula C8H12N2O4S2
Molecular Weight (g/mol) 264.31
MDL Number MFCD00276605
InChI Key BYZJRLRCNOECEV-UHFFFAOYSA-N
Synonym 1,2-bis methanesulfonamido benzene, n,n'-1,2-phenylene dimethanesulfonamide, n-2-methanesulfonamidophenyl methanesulfonamide, n-2-methylsulfonyl amino phenyl methanesulfonamide, methylsulfonyl 2-methylsulfonyl amino phenyl amine, maybridge1_000065, n,n'-o-phenylenebis methanesulfonamide, o-phenylenediamine n,n'-bis methanesulfonamide, n-2-methanesulfonamido phenyl methanesulfonamide
PubChem CID 227307
IUPAC Name N-[2-(methanesulfonamido)phenyl]methanesulfonamide
SMILES CS(=O)(=O)NC1=CC=CC=C1NS(C)(=O)=O

Specifications

Melting Point 211°C to 214°C
Odor Odorless
Quantity 2.5 g
Formula Weight 264.32
Percent Purity 97%
Chemical Name or Material 1,2-Bis(methanesulfonamido)benzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.