missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(phenylsulfonyl)ethane, 98+%

Product Code. 11316786
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11316786 25 g 25g
11306786 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11316786 Supplier Thermo Scientific Alfa Aesar Supplier No. L04083.14
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 599-94-0 | C14H14O4S2 | 310.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 599-94-0
Molecular Formula C14H14O4S2
Molecular Weight (g/mol) 310.38
MDL Number MFCD00041256
InChI Key ULELOBVZIKJPAC-UHFFFAOYSA-N
Synonym 1,2-bis phenylsulfonyl ethane, 1,2-bis phenylsulphonyl ethane, 1,1'-ethane-1,2-diylbis sulphonyl bisbenzene, 2-benzenesulfonyl ethylsulfonylbenzene, ethylenebisphenyl sulfone, 2-phenylsulfonyl ethyl sulfonyl benzene, 2-benzenesulfonyl ethanesulfonyl benzene, 1,1'-ethane-1,2-diyldisulfonyl dibenzene, ethylene diphenylsulfone, acmc-1apw3
PubChem CID 69036
IUPAC Name 2-(benzenesulfonyl)ethylsulfonylbenzene
SMILES O=S(=O)(CCS(=O)(=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 179°C to 180°C
Quantity 25 g
Beilstein 1888062
Formula Weight 310.39
Percent Purity ≥98%
Chemical Name or Material 1,2-Bis(phenylsulfonyl)ethane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.