missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Bromoethoxy)-3-nitrobenzene, 97%, Thermo Scientific™

Product Code. 10651885 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10651885 25 g Glass bottle 25g
10045982 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10651885 Supplier Thermo Scientific Acros Supplier No. 385260250
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 13831-59-9
Molecular Formula C8H8BrNO3
Molecular Weight (g/mol) 246.06
MDL Number MFCD07783648
InChI Key QBUSKXLDUNPEMZ-UHFFFAOYSA-N
Synonym 1-2-bromoethoxy-3-nitrobenzene, benzene,1-2-bromoethoxy-3-nitro, 2-bromoethyl 3-nitrophenyl ether, acmc-209chl, 2-bromo-3'-nitrophenetole, 2-3-nitrophenoxy ethyl bromide, 3-2-bromoethoxy-1-nitrobenzene
PubChem CID 269626
IUPAC Name 1-(2-bromoethoxy)-3-nitrobenzene
SMILES [O-][N+](=O)C1=CC=CC(OCCBr)=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 39°C
Color Brown or Green-Yellow
Boiling Point 140°C to 145°C (2.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula NO2C6H4OCH2CH2Br
Refractive Index 1.5830 to 1.585
Quantity 25 g
Formula Weight 246.06
Percent Purity 97%
Physical Form Low Melting Solid
Chemical Name or Material 1-(2-Bromoethoxy)-3-nitrobenzene
Show More Show Less
compliance-icons
Product Identifier
  • 1-(2-Bromoethoxy)-3-nitrobenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.