missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Bromoethoxy)-3-nitrobenzene, 97%, Thermo Scientific™

Product Code. p-3211163 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10045982

Brand: Thermo Scientific Acros 385260050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 13831-59-9
Molecular Formula C8H8BrNO3
Molecular Weight (g/mol) 246.06
MDL Number MFCD07783648
InChI Key QBUSKXLDUNPEMZ-UHFFFAOYSA-N
Synonym 1-2-bromoethoxy-3-nitrobenzene, benzene,1-2-bromoethoxy-3-nitro, 2-bromoethyl 3-nitrophenyl ether, acmc-209chl, 2-bromo-3'-nitrophenetole, 2-3-nitrophenoxy ethyl bromide, 3-2-bromoethoxy-1-nitrobenzene
PubChem CID 269626
IUPAC Name 1-(2-bromoethoxy)-3-nitrobenzene
SMILES [O-][N+](=O)C1=CC=CC(OCCBr)=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 39°C
Color Brown or Green-Yellow
Boiling Point 140°C to 145°C (2.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula NO2C6H4OCH2CH2Br
Refractive Index 1.5830 to 1.585
Quantity 5 g
Formula Weight 246.06
Percent Purity 97%
Physical Form Low Melting Solid
Chemical Name or Material 1-(2-Bromoethoxy)-3-nitrobenzene
Show More Show Less
compliance-icons
Product Identifier
  • 1-(2-Bromoethoxy)-3-nitrobenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.