missing translation for 'onlineSavingsMsg'
Learn More

1-[2-Chloro-5-(trifluoromethyl)phenyl]hydrazine, ≥95%, Thermo Scientific™

Product Code. 10477812
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10477812 1 g Amber Glass Bottle 1g
10252312 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10477812 Supplier Thermo Scientific Maybridge Supplier No. RF01368DA
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1869-22-3
Molecular Formula C7H6ClF3N2
Molecular Weight (g/mol) 210.584
MDL Number MFCD00052685
InChI Key PAPSYPXQNRTIRR-UHFFFAOYSA-N
Synonym 1-2-chloro-5-trifluoromethyl phenyl hydrazine, 2-chloro-5-trifluoromethyl phenyl hydrazine, 2-chloro-5-trifluoromethyl phenylhydrazine, hydrazine, 2-chloro-5-trifluoromethyl phenyl, 4-chloro-3-hydrazinobenzotrifluoride, 2-chloro-5-trifluoromethylphenylhydrazine
PubChem CID 2060249
IUPAC Name [2-chloro-5-(trifluoromethyl)phenyl]hydrazine
SMILES C1=CC(=C(C=C1C(F)(F)F)NN)Cl

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 210.59
Percent Purity 95%
Chemical Name or Material 1-[2-Chloro-5-(trifluoromethyl)phenyl]hydrazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.