missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Chloroethyl)-4-fluorobenzene, 97%, Thermo Scientific™

Product Code. p-7046897
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11389355

Brand: Thermo Scientific Alfa Aesar L16719.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 332-43-4
Molecular Formula C8H8ClF
Molecular Weight (g/mol) 158.60
MDL Number MFCD00155000
InChI Key LPUURRVXQCVXCX-UHFFFAOYSA-N
Synonym 1-2-chloroethyl-4-fluorobenzene, 4-fluorophenethyl chloride, 2-4-fluorophenyl ethyl chloride, 1-2'-chloroethyl-4-fluorobenzene, 1-2-chloro-ethyl-4-fluoro-benzene, benzene, 1-2-chloroethyl-4-fluoro, acmc-1anql, 4-fluorophenetyl chloride, ksc570e7d, 1-fluoro-4-2-chloroethyl benzene
PubChem CID 67622
IUPAC Name 1-(2-chloroethyl)-4-fluorobenzene
SMILES FC1=CC=C(CCCl)C=C1

Specifications

Density 1.507
Boiling Point 98°C to 99°C (26 mmHg)
Refractive Index 1.508
Quantity 1 g
Beilstein 2079481
Formula Weight 158.6
Percent Purity 97%
Chemical Name or Material 1-(2-Chloroethyl)-4-fluorobenzene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.