missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Chloroethyl)-4-methoxybenzene, 97%, Thermo Scientific Chemicals

Product Code. 10757772 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
5mL
This item is not returnable. View return policy

Product Code. 10757772

Brand: Thermo Scientific Acros 123110050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18217-00-0
Molecular Formula C9H11ClO
Molecular Weight (g/mol) 170.64
MDL Number MFCD00044718
InChI Key PMIAMRAWHYEPNH-UHFFFAOYSA-N
Synonym 1-2-chloroethyl-4-methoxybenzene, 4-2-chloroethyl anisole, 4-2-chloroethyl phenyl methyl ether, benzene, 1-2-chloroethyl-4-methoxy, p-2-chloro ethyl anisole, 2-4-methoxyphenyl-ethylchloride, 4-methoxyphenethyl chloride, acmc-1c78x, 4-methoxyphenylethyl chloride, 2-4-methoxyphenyl ethylchloride
PubChem CID 87513
IUPAC Name 1-(2-chloroethyl)-4-methoxybenzene
SMILES COC1=CC=C(CCCl)C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1300g/mL
Boiling Point 125°C to 128°C (13.0 mbar)
Flash Point 122°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H4(CH2)2Cl
Refractive Index 1.5360 to 1.538
Quantity 5 mL
Specific Gravity 1.13
Solubility Information Solubility in water: 130mg/l (20°C). Other solubilities: soluble in methanol,ethanol,2-propanol,,n,n-dimethylformamide,acetone and dichloromethane
Formula Weight 170.64
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-(2-Chloroethyl)-4-methoxybenzene, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-(2-Chloroethyl)-4-methoxybenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.