missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dehydro Aldosterone 21-Acetate, TRC

Product Code. 30352343
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
2.5 mg
Unit Size:
0.5mg
1mg
2.50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30352343 2.5 mg 2.50mg
30352486 0.5 mg 0.5mg
30359607 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30352343 Supplier Toronto Research Chemicals Supplier No. TRCD2291082.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2-Dehydro Aldosterone 21-Acetate
Analyte or Component Names 1,2-Dehydro Aldosterone 21-Acetate
Shipping Condition Room Temperature
Molecular Formula C23 H28 O6
InChI Formula InChI=1S/C23H28O6/c1-12(24)28-11-18(26)17-6-5-16-15-4-3-13-9-14(25)7-8-22(13,2)20(15)19-10-23(16,17)21(27)29-19/h7-9,15-17,19-21,27H,3-6,10-11H2,1-2H3/t15-,16-,17+,19-,20+,21?,22+,23+/m1/s1
Solution Type Neat
SMILES CC(=O)OCC(=O)[C@@H]1CC[C@@H]2[C@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@H]3[C@H]5C[C@@]12C(O)O5
IUPAC Name [2-[(1S,2R,5R,6R,14R,15R,16R)-18-hydroxy-14-methyl-11-oxo-17-oxapentacyclo[14.2.1.01,5.06,15.09,14]nonadeca-9,12-dien-2-yl]-2-oxoethyl] acetate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 400.465
Quantity 2.5 mg
Formula Weight 400.189
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.