missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromobenzene, 98%

Product Code. 10489232 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10489232 25 mL Glass bottle 25mL
15780859 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10489232 Supplier Thermo Scientific Acros Supplier No. 348930250
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 583-53-9 | C6H4Br2 | 235.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 583-53-9
Molecular Formula C6H4Br2
Molecular Weight (g/mol) 235.91
MDL Number MFCD00000057
InChI Key WQONPSCCEXUXTQ-UHFFFAOYSA-N
Synonym o-dibromobenzene, dibromobenzene, benzene, 1,2-dibromo, ortho-dibromobenzene, benzene, o-dibromo, benzene, dibromo, unii-52k9w7u6eh, dibromobenzene mixed isomers, 1,2 dibromobenzene, 1,6-dibromobenzene
PubChem CID 11414
ChEBI CHEBI:37152
IUPAC Name 1,2-dibromobenzene
SMILES C1=CC=C(C(=C1)Br)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 4.0°C to 6.0°C
Color Colorless to Yellow or Pink
Density 1.9560g/mL
Boiling Point 224.0°C
Flash Point 91°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H4Br2
Refractive Index 1.6100 to 1.6120
Quantity 25 mL
Beilstein 05, 210
Specific Gravity 1.956
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol,acetone,ether,benzene,ca,rbon tetrachloride,n-heptane
Formula Weight 235.91
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1, 2-Dibromobenzene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,2-Dibromobenzene
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.