missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloro-4-iodobenzene, 98%

Product Code. 11435493
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11435493 50 g 50g
11425493 10 g 10g
11445493 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11435493 Supplier Thermo Scientific Alfa Aesar Supplier No. A13440.18
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 20555-91-3 | C6H3Cl2I | 272.894 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20555-91-3
Molecular Formula C6H3Cl2I
Molecular Weight (g/mol) 272.894
MDL Number MFCD00019014
InChI Key NADPFZNWCQIJJW-UHFFFAOYSA-N
Synonym 3,4-dichloroiodobenzene, benzene, 1,2-dichloro-4-iodo, maybridge1_001437, pubchem3691, acmc-209fbv, 3,4-dichloro-iodobenzene, 1,2-dichloro-5-iodobenzene, 1-iodo-3,4-dichlorobenzene, 3,4-dichloro-1-iodobenzene, 4-iodo-1,2-dichlorobenzene
PubChem CID 88591
IUPAC Name 1,2-dichloro-4-iodobenzene
SMILES C1=CC(=C(C=C1I)Cl)Cl

Specifications

Melting Point 29°C to 31°C
Boiling Point 116°C to 118°C (12 mmHg)
Refractive Index 1.648
Quantity 50 g
Beilstein 2245136
Sensitivity Light sensitive
Formula Weight 272.9
Percent Purity 98%
Chemical Name or Material 1,2-Dichloro-4-iodobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.