missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloropropane, 98%

Product Code. 11392016
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11392016 25 g 25g
11302026 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392016 Supplier Thermo Scientific Alfa Aesar Supplier No. L03083.14
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 78-87-5 | C3H6Cl2 | 112.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-87-5
Molecular Formula C3H6Cl2
Molecular Weight (g/mol) 112.98
MDL Number MFCD00000868
InChI Key KNKRKFALVUDBJE-UHFFFAOYNA-N
Synonym propylene dichloride, propane, 1,2-dichloro, propylene chloride, dwuchloropropan, bichlorure de propylene, rcra waste number u083, caswell no. 324, dwuchloropropan polish, alpha,beta-dichloropropane, ccris 951
PubChem CID 6564
ChEBI CHEBI:82163
IUPAC Name 1,2-dichloropropane
SMILES CC(Cl)CCl

Specifications

Melting Point -100°C
Density 1.157
Boiling Point 95°C to 97°C
Flash Point 4°C (39°F)
Odor Sweet
Refractive Index 1.438
Quantity 25 g
UN Number UN1279
Beilstein 1718880
Merck Index 14,7854
Formula Weight 112.99
Percent Purity 98%
Chemical Name or Material 1,2-Dichloropropane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.