accessibility menu, dialog, popup

UserName

CH3ClCl

CAS RN 78-87-5

IUPAC Name: 1,2-dichloropropane
Synonyms: 1,2-dichloropropane caswell no. 324 epa pesticide chemical code 029002 dichloro-1,2 propane C3H6cl2 dwuchloropropan ccris 951 rcra waste number U083 bichlorure de propylene
Molecular Weight (g/mol): 112.98
Molecular Formula: C3H6Cl2
InChi Key: KNKRKFALVUDBJE-UHFFFAOYNA-N
SMILES: CC(Cl)CCl
Molecular Weight (g/mol) 
  • (9)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (4)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (9)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (4)
  • (4)
  • (2)
  • (1)
  • (2)