missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloropropane, TRC

Product Code. 30135213
Change view
Click to view available options
Quantity:
1 g
100 g
Unit Size:
100g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135213 100 g 100g
30010591 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135213 Supplier Toronto Research Chemicals Supplier No. TRCD434520100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-87-5
Molecular Formula C3 H6 Cl2
Molecular Weight (g/mol) 112.99
Synonym 1,2-Dichloropropane, 1,2-DCP, NSC 1237, Propylene chloride, Propylene dichloride, R 270da
IUPAC Name 1,2-dichloropropane
SMILES CC(Cl)CCl

Specifications

Quantity 100 g
InChI Formula InChI=1S/C3H6Cl2/c1-3(5)2-4/h3H,2H2,1H3
Formula Weight 111.9847
Chemical Name or Material 1,2-Dichloropropane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.