missing translation for 'onlineSavingsMsg'
Learn More

1,2-difluoro-4-iodobenzene, Thermo Scientific™

Product Code. 10460754
Change view
Click to view available options
Quantity:
100 g
Packaging:
Amber Glass Bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10460754 100 g Amber Glass Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10460754 Supplier Thermo Scientific Maybridge Supplier No. SB01245FA
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 64248-58-4
Density 1.9900g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C6H3F2I
Linear Formula IC6H3F2
MDL Number MFCD00044630
Quantity 100 g
Synonym 3,4-difluoroiodobenzene, 1,2-difluoro-4-iodo-benzene, benzene, 1,2-difluoro-4-iodo, 3,4-difluoro iodobenzene, pubchem3461, 3,4-difluoro-iodobenzene, 3,4-difluorophenyl iodide, 3,4-difluoro-1-iodobenzene, 1-iodo-3,4-difluoro-benzene
InChI Key KSASJELKLBIMSG-UHFFFAOYSA-N
SMILES FC1=C(F)C=C(I)C=C1
IUPAC Name 1,2-difluoro-4-iodobenzene
Molecular Weight (g/mol) 239.99
PubChem CID 2724444
Formula Weight 239.99
Percent Purity 97%
Chemical Name or Material 1, 2-Difluoro-4-iodobenzene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.