missing translation for 'onlineSavingsMsg'
Learn More

1,2-Difluorobenzene, 98%

Product Code. 10724271 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Unit Size:
10mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10724271 10 mL Glass bottle 10mL
10551211 50 mL Glass bottle 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10724271 Supplier Thermo Scientific Acros Supplier No. 155150100
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 367-11-3 | C6H4F2 | 114.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 367-11-3
Molecular Formula C6H4F2
Molecular Weight (g/mol) 114.10
MDL Number MFCD00000284
InChI Key GOYDNIKZWGIXJT-UHFFFAOYSA-N
Synonym o-difluorobenzene, benzene, 1,2-difluoro, benzene, o-difluoro, ortho-difluorobenzene, o-difluoro benzene, difluorobenzene, 1,2-difluoro-benzene, unii-aw7qgmw29c, aw7qgmw29c, 0-difluorobenzene
PubChem CID 9706
ChEBI CHEBI:38583
IUPAC Name 1,2-difluorobenzene
SMILES FC1=CC=CC=C1F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -34°C
Color Colorless
Density 1.1580g/mL
Boiling Point 92°C
Flash Point 2°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C6H4F2
Refractive Index 1.4417 to 1.4437
Quantity 10 mL
Beilstein 05, IV, 637
Specific Gravity 1.158
Formula Weight 114.09
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1, 2-Difluorobenzene, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 1,2-Difluorobenzene
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.